missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAEP/Glycodelin Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £349.00
Specifications
| Antigen | PAEP/Glycodelin |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236333
|
Proteintech
18187-1-AP-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236656
|
Proteintech
18187-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PAEP/Glycodelin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PAEP/Glycodelin. This antibody recognizes human antigen. The PAEP/Glycodelin antibody has been validated for the following applications: IHC, ELISA analysis.Specifications
| PAEP/Glycodelin | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| GD; GdA; GdF; GdS; MGC138509; MGC142288; PAEG; PEP; PP14 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 300 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 5047 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title