missing translation for 'onlineSavingsMsg'
Learn More

Paeonol, MedChemExpress

Product Code. 16432509
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432509 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432509 Supplier Medchem Express Supplier No. HYN0159
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Paeonol is an active extraction from the root of Paeonia suffruticosa, Paeonol inhibits MAO-A and MAO-B with IC50 of 54.6 μM and 42.5 μM, respectively.

Paeonol is an active extraction from the root of Paeonia suffruticosa, Paeonol inhibits MAO-A and MAO-B with IC50 of 54.6 μM and 42.5 μM, respectively.

Specifications

Chemical Name or Material Paeonol
CAS 552-41-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H10O3
Quantity 1 g
Solubility Information Ethanol : 50 mg/mL (300.90 mM; Need ultrasonic) ∣DMSO : ≥ 38 mg/mL (228.68 mM) ∣H2O : 1 mg/mL (6.02 mM; Need ultrasonic)
SMILES CC(C1=CC=C(OC)C=C1O)=O
Molecular Weight (g/mol) 166.17
Formula Weight 166.17
Percent Purity 99.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.