missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PADI4 Monoclonal Antibody (OTI6A2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526006
This item is not returnable.
View return policy
Description
PADI4 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow
Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at 'Arg-8' and/or 'Arg-17' and histone H4 at 'Arg-3', which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at 'Arg-2142', which favors its interaction with NCOA2/GRIP1.
Specifications
| PADI4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UM07 | |
| Padi4 | |
| Human recombinant protein fragment corresponding to amnio acids 299-588 of PADI4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI6A2 | |
| Unconjugated | |
| Padi4 | |
| HL-60 PAD; PAD; PAD type IV; PAD4; Padi4; PADI5; PADI-H protein; PAD-R4; Pdi4; PDI5; peptidyl arginine deiminase 4; peptidyl arginine deiminase, type 4; peptidyl arginine deiminase, type IV; peptidyl arginine deiminase, type V; peptidylarginine deiminase IV; Peptidylarginine deiminase type alpha; protein-arginine deiminase type IV; Protein-arginine deiminase type-4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23569 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction