missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PADI4 Monoclonal antibody specifically detects PADI4 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | PADI4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI4H5 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human PADI4(NP_036519) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?