missing translation for 'onlineSavingsMsg'
Learn More

PADI4 Antibody, HRP, Novus Biologicals™

Product Code. 18382756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382756 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382756 Supplier Novus Biologicals Supplier No. NBP299694H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PADI4 Polyclonal specifically detects PADI4 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI4
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant human PADI4 (Met1-Pro663).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23569
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.