missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PADI4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£176.00 - £367.00
Specifications
| Antigen | PADI4 |
|---|---|
| Concentration | 2.2 mg/mL |
| Dilution | Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative), ICC/IF (Negative) |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18034057
|
Novus Biologicals
NBP1-97309SS |
0.025 mL |
£176.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745233
|
Novus Biologicals
NBP1-97309 |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PADI4 Polyclonal specifically detects PADI4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative).Specifications
| PADI4 | |
| Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative), ICC/IF (Negative) | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.4) with 0.02% Sodium Azide | |
| HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4 | |
| PADI4 | |
| IgG | |
| Affinity Purified |
| 2.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Mouse | |
| Q9UM07 | |
| 23569 | |
| A synthetic peptide made to an internal portion of the human PAD4 protein (between residues 200-250) [UniProt Q9UM07] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title