missing translation for 'onlineSavingsMsg'
Learn More

PADI2 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30731589 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731589 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731589 Supplier Novus Biologicals Supplier No. AF7257Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PADI2 Polyclonal antibody specifically detects PADI2 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.3.15, KIAA0994PDI2PAD2, PAD-H19, peptidlyarginine deiminase type II, peptidyl arginine deiminase, type II, Peptidylarginine deiminase II, protein arginine deiminase, Protein-arginine deiminase type II, protein-arginine deiminase type-2
Host Species Sheep
Immunogen E. coli-derived recombinant human PADI2, Met1-Ala115, Accession # Q9Y2J8
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Amino Acids Drugs and other small molecules, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11240
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.