missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PADI2 Antibody (4D4), Novus Biologicals™

Product Code. 18344689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344689 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344689 Supplier Novus Biologicals™ Supplier No. H00011240M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PADI2 Monoclonal antibody specifically detects PADI2 in Human,Mouse samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI2
Applications Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA
Classification Monoclonal
Clone 4D4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_031391.2
Gene Alias EC 3.5.3.15, KIAA0994PDI2PAD2, PAD-H19, peptidlyarginine deiminase type II, peptidyl arginine deiminase, type II, Peptidylarginine deiminase II, protein arginine deiminase, Protein-arginine deiminase type II, protein-arginine deiminase type-2
Host Species Mouse
Immunogen PADI2 (NP_003008, 1 a.a. ∽ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLRERTVRLQYGSRVEAVYVLGTYLWTDVYSAAPAGAQTFSLKHSEHVWVEVVRDGEAEEVATNGKQRWLLSPSTTLRVTMSQASTEASSDKVTVNYYDEEGSIPIDQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Amino Acids Drugs and other small molecules, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11240
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.