missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACT Recombinant Rabbit Monoclonal Antibody (23GB2340), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554095
This item is not returnable.
View return policy
Description
PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
Specifications
| PACT | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRKRA | |
| AV120107; DYT16; HSD14; HSD-14; interferon-inducible double stranded RNA-dependent protein kinase activator A; interferon-inducible double-stranded RNA-dependent protein kinase activator A; lear; PACT; PKR associated protein X; PKR-associated protein X; PKR-associating protein X; PRK; PRKRA; protein activator of interferon induced protein kinase EIF2AK2; protein activator of the interferon-induced protein kinase; protein kinase, interferon inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23992, 311130, 8575 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2340 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75569, Q4V8C7, Q9WTX2 | |
| PRKRA | |
| A synthesized peptide derived from human PRKRA (260-313AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering