missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACT Rabbit anti-Human, Mouse, Rat, Clone: S04-8F7, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | PACT |
|---|---|
| Clone | S04-8F7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18325784
|
Novus Biologicals
NBP3-15072-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18365174
|
Novus Biologicals
NBP3-15072-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PACT Monoclonal antibody specifically detects PACT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifica
| PACT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 8575 | |
| IgG | |
| Affinity-purified |
| S04-8F7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DYT16interferon-inducible double stranded RNA-dependent activator, PACTPKR-associating protein X, PKR-associated protein X, protein kinase, interferon-inducible double stranded RNA dependent activator, RAXHSD14 | |
| A synthetic peptide of of human PACT (Uniprot#: O75569) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto