missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACT Rabbit anti-Human, Mouse, Rat, Clone: 2W2E10, Novus Biologicals™
Rabbit Monoclonal Antibody
£163.00 - £386.00
Specifications
| Antigen | PACT |
|---|---|
| Clone | 2W2E10 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18355756
|
Novus Biologicals
NBP3-16701-20UL |
20 μg |
£163.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18380464
|
Novus Biologicals
NBP3-16701-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PACT Monoclonal antibody specifically detects PACT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PACT | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DYT16interferon-inducible double stranded RNA-dependent activator, PACTPKR-associating protein X, PKR-associated protein X, protein kinase, interferon-inducible double stranded RNA dependent activator, RAXHSD14 | |
| A synthetic peptide corresponding to a sequence within amino acids 214-313 of human PACT (O75569). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 2W2E10 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8575 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title