missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102768
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Antibody detects endogenous levels of total PRKRA.
PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
Specifikationer
| PACT | |
| Polyclonal | |
| Unconjugated | |
| PRKRA | |
| AV120107; DYT16; HSD14; HSD-14; interferon-inducible double stranded RNA-dependent protein kinase activator A; interferon-inducible double-stranded RNA-dependent protein kinase activator A; lear; PACT; PKR associated protein X; PKR-associated protein X; PKR-associating protein X; PRK; PRKRA; protein activator of interferon induced protein kinase EIF2AK2; protein activator of the interferon-induced protein kinase; protein kinase, interferon inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23992, 311130, 8575 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75569, Q4V8C7, Q9WTX2 | |
| PRKRA | |
| A synthesized peptide derived from human PRKRA(Accession O75569), corresponding to amino acid residues A94-E144. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering