missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529950
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, Jurkat, NT2D. Predicted reactivity: Mouse (98%), Rat (98%), Rhesus Monkey (100%), Chimpanzee (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
Specifications
| PACT | |
| Polyclonal | |
| Unconjugated | |
| PRKRA | |
| AV120107; DYT16; HSD14; HSD-14; interferon-inducible double stranded RNA-dependent protein kinase activator A; interferon-inducible double-stranded RNA-dependent protein kinase activator A; lear; PACT; PKR associated protein X; PKR-associated protein X; PKR-associating protein X; PRK; PRKRA; protein activator of interferon induced protein kinase EIF2AK2; protein activator of the interferon-induced protein kinase; protein kinase, interferon inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8575 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.51 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75569 | |
| PRKRA | |
| Recombinant protein encompassing a sequence within the center region of human PACT. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Amphibian, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction