missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACSIN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583465
This item is not returnable.
View return policy
Description
Immunogen sequence: EWSLDTQRTI SRKEKGGRSP DEVTLTSIVP TRDGTAPPPQ SPGSPGTGQD EEWSDEESP Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.
This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.
Specifications
| PACSIN3 | |
| Polyclonal | |
| Unconjugated | |
| PACSIN3 | |
| 4921507A02Rik; 6330413E15Rik; AW413130; EGK_06300; Endophilin I; PACSIN3; Protein kinase C and casein kinase II substrate protein 3; protein kinase C and casein kinase substrate in neurons 3; protein kinase C and casein kinase substrate in neurons protein 3; SDPIII; SH3 domain-containing protein 6511; syndapin III | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKS6 | |
| PACSIN3 | |
| Recombinant protein corresponding to Human PACSIN3. Recombinant protein control fragment (Product #RP-97429). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction