missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACSIN3 Monoclonal Antibody (OTI4F8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525377
This item is not returnable.
View return policy
Description
PACSIN3 Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.
Specifications
| PACSIN3 | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UKS6 | |
| PACSIN3 | |
| Full length human recombinant protein of PACSIN3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F8 | |
| Unconjugated | |
| PACSIN3 | |
| 4921507A02Rik; 6330413E15Rik; AW413130; EGK_06300; Endophilin I; PACSIN3; Protein kinase C and casein kinase II substrate protein 3; protein kinase C and casein kinase substrate in neurons 3; protein kinase C and casein kinase substrate in neurons protein 3; SDPIII; SH3 domain-containing protein 6511; syndapin III | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29763 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction