missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACSIN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38573-20ul
This item is not returnable.
View return policy
Description
PACSIN3 Polyclonal antibody specifically detects PACSIN3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PACSIN3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Endophilin I, FLJ17861, protein kinase C and casein kinase substrate in neurons 3, protein kinase C and casein kinase substrate in neurons protein 3, SDPIII, SH3 domain-containing protein 6511, syndapin III | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 175-424 of human PACSIN3 (NP_057307.2).,, Sequence:, KADSAVSQEQLRKLQERVERCAKEAEKTKAQYEQTLAELHRYTPRYMEDMEQAFETCQAAERQRLLFFKDMLLTLHQHLDLSSSEKFHELHRDLHQGIEAASDEEDLRWWRSTHGPGMAMNWPQFEEWSLDTQRTISRKEKGGRSPDEVTLTSIVPTRDGTAPPPQSPGSPGTGQDEEWSDEESPRKAATGVRVRALYDYAGQEADELSFRAGEELLKMSEEDEQGWCQGQLQSGRIGLYPANYVECVGA | |
| 20 μL | |
| Cancer, Diabetes Research, Neuroscience, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| 29763 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction