missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PACSIN1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA584257

Product Code. 16301215

  • £452.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EKQPKKAEGV ALTNATGAVE STSQAGDRGS VSSYDRGQPY ATEWSDDESG N Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 94%.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PACSIN1
Polyclonal
Unconjugated
Pacsin1
A830061D09Rik; Dynamin PRD-interacting protein; dynamin proline-rich domain-interacting protein; H74; KIAA1379; mKIAA1379; Pacsin; Pacsin1; protein kinase C and casein kinase substrate in neurons 1; protein kinase C and casein kinase substrate in neurons protein 1; SdpI; synaptic, dynamin-associated protein I; syndapin; syndapin 1; syndapin I; syndapin-1; Syndapin-I
Rabbit
Antigen affinity chromatography
RUO
29993
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BY11
Pacsin1
Recombinant protein corresponding to Human PACSIN1. Recombinant protein control fragment (Product #RP-101745).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.