missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACSIN1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA517994
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
| PACSIN1 | |
| Polyclonal | |
| Unconjugated | |
| Pacsin1 | |
| A830061D09Rik; Dynamin PRD-interacting protein; dynamin proline-rich domain-interacting protein; H74; KIAA1379; mKIAA1379; Pacsin; Pacsin1; protein kinase C and casein kinase substrate in neurons 1; protein kinase C and casein kinase substrate in neurons protein 1; SdpI; synaptic, dynamin-associated protein I; syndapin; syndapin 1; syndapin I; syndapin-1; Syndapin-I | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 29993 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BY11 | |
| Pacsin1 | |
| Synthetic peptide sequence (SSSYDEASLAPEET) corresponding to the N-terminus amino acids of PACSIN1 (aa 2-16). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction