missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACS1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | PACS1 |
|---|---|
| Dilution | Western Blot 1:1000-1:3000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PACS1 Polyclonal antibody specifically detects PACS1 in Human samples. It is validated for Western BlotSpecifications
| PACS1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, metabolism, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 55690 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:3000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cytosolic sorting protein PACS-1, FLJ10209, KIAA1175, PACS-1, phosphofurin acidic cluster sorting protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 420-560 of human PACS1 (NP_060496.2). SKGSLGKDTTSPMELAALEKIKSTWIKNQDDSLTETDTLEITDQDMFGDASTSLVVPEKVKTPMKSSKTDLQGSASPSKVEGVHTPRQKRSTPLKERQLSKPLSERTNSSDSERSPDLGHSTQIPRKVVYDQLNQILVSDA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title