missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACRGL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569871
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
PACRGL (Parkin Coregulated Like) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includebinding.
Specifications
| PACRGL | |
| Polyclonal | |
| Unconjugated | |
| PACRGL | |
| 4933428G09Rik; C4orf28; Pacrgl; PACRG-like protein; PARK2 coregulated like; PARK2 co-regulated like; PARK2 co-regulated-like; parkin coregulated like; RGD1311045 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 133015 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N7B6 | |
| PACRGL | |
| synthetic peptide directed towards the middle region of human PACRGL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction