Learn More
PACRG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543934
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GAIMARCNLD HLGSSDPPTS ASQVAEIIVN SGDGIDYSQQ KRENIGDLIQ Sequence homology: Cow: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
PACRG Polyclonal antibody specifically detects PACRG in Human samples. It is validated for Western Blot.Specifications
PACRG | |
Polyclonal | |
Unconjugated | |
PACRG | |
1700008H23Rik; FLJ32724; GLUP; HAK005771; Hypertension-related protein 1-like protein; molecular chaperone/chaperonin-binding protein; PACRG; PARK2 coregulated; PARK2 co-regulated; PARK2 coregulated gene protein; PARK2CRG; parkin coregulated; parkin coregulated gene protein; parkin co-regulated gene protein; Parkin coregulated gene protein homolog; RGD1561027; RP3-495O10.2; Unknown (protein for MGC:134579); zgc:101786 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
135138 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96M98 | |
PACRG | |
Synthetic peptide directed towards the middle region of human PACRG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |