missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACRG Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566834
This item is not returnable.
View return policy
Description
Immunogen sequence: AFKERPTKPTA FRKFYERGDF PIALEHDSKG NKIAWKVEIE KLDYHHYLPL F Highest antigen sequence identity to the following orthologs - mouse 94%, rat 29%.
PACRG is a protein that is conserved across metazoans. In vertebrates, this gene is linked in a head-to-head arrangement with the adjacent parkin gene, which is associated with autosomal recessive juvenile Parkinson's disease. These genes are co-regulated in various tissues and they share a bi-directional promoter. Both genes are associated with susceptibility to leprosy. The parkin co-regulated gene protein forms a large molecular complex with chaperones, including heat shock proteins 70 and 90, and chaperonin components. This protein is also a component of Lewy bodies in Parkinson's disease patients, and it suppresses unfolded Pael receptor-induced neuronal cell death.
Specifications
| PACRG | |
| Polyclonal | |
| Unconjugated | |
| PACRG | |
| 1700008H23Rik; FLJ32724; GLUP; HAK005771; Hypertension-related protein 1-like protein; molecular chaperone/chaperonin-binding protein; PACRG; PARK2 coregulated; PARK2 co-regulated; PARK2 coregulated gene protein; PARK2CRG; parkin coregulated; parkin coregulated gene protein; parkin co-regulated gene protein; Parkin coregulated gene protein homolog; RGD1561027; RP3-495O10.2; Unknown (protein for MGC:134579); zgc:101786 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 135138 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96M98 | |
| PACRG | |
| Recombinant Human PACRG. Recombinant protein control fragment (Product #RP-107490). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction