missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACRG Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | PACRG |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PACRG Polyclonal antibody specifically detects PACRG in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PACRG | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 135138 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ32724, Glup, HAK005771, Molecular chaperone/chaperonin-binding protein, PARK2 co-regulated, PARK2 coregulated gene protein, PARK2CRG, parkin coregulated gene protein, parkin co-regulated gene protein, RP3-495O10.2 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human PACRG (NP_001073847.1). IIPIKNALNLRNRQVICVTLKVLQHLVVSAEMVGKALVPYYRQILPVLNIFKNMNVNSGDGIDYSQQKRENIGDLIQETLEAFERYGGENAFINIKYVVPT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title