missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PACMA 31

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PACMA 31 is an irreversible protein disulfide isomerase (PDI) inhibitor; inhibits growth of ovarian cancer cells. Suppresses tumor growth in a mouse xenograph model of human ovarian cancer.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | PACMA 31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 430.47 |
| Inhibitors | PACMA 31 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction