missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PACMA 31
GREENER_CHOICE

Product Code. 18778048
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18778048

Brand: Tocris Bioscience™ 5116/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Irreversible protein disulfide isomerase (PDI) inhibitor

PACMA 31 is an irreversible protein disulfide isomerase (PDI) inhibitor; inhibits growth of ovarian cancer cells. Suppresses tumor growth in a mouse xenograph model of human ovarian cancer.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type PACMA 31
Purity >99%
Molecular Weight (g/mol) 430.47
Inhibitors PACMA 31

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.