missing translation for 'onlineSavingsMsg'
Learn More

Paclitaxel, TRC

Product Code. 30410685
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410685 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410685 Supplier Toronto Research Chemicals Supplier No. TRCP13250025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paclitaxel
CAS 33069-62-4
Purity Grade Notes HPLC
Molecular Formula C47 H51 N O14
Quantity 25 mg
Synonym Paclitaxel, 5β,20-Epoxy-1,7β-dihydroxy-9-oxotax-11-ene-2α,4,10β,13α-tetrayl 4,10-diacetate 2-benzoate 13-[(2R,3S)-3-(benzoylamino)-2-hydroxy-3-phenylpropanoate], Benzenepropanoic acid','β-(benzoylamino)-α-hydroxy-','(2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester','(αR,βS)-, Benzenepropanoic acid','β-(benzoylamino)-α-hydroxy-','6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester','[2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, Tax-11-en-9-one','5β,20-epoxy-1,2α,4,7β,10β,13α-hexahydroxy-','4,10-diacetate 2-benzoate 13-ester with (2R,3S)-N-benzoyl-3-phenylisoserine (8CI), 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete','benzenepropanoic acid deriv., (-)-Paclitaxel, 5β,20-Epoxy-1,2a,4,7β,10β,13α-hexahydroxy-tax-11-en-9-one 4,10-diacetate 2-benzoate 13-ester with (2R,3S)-N-benzoyl-3-phenylisoserine, ABI 007, Abraxane, Anzatax, BMS 181339-01, Capxol, Coroflex Please, Cremophor-free paclitaxel, Cyclopax, Cynviloq, Cypher select, DHP 107, Ebetaxel, EmPAC, EndoTAG 1, Genaxol, Genetaxyl, Genexol, Genexol-PM, IG 001, IG 002, Infinnium, Intaxel, LEP-ETU, Liposome-entrapped paclitaxel easy-to-use, MBT 0206, Mitotax, NK 105, NSC 125973, Nab-paclitaxel, Nanotax, Nanoxel, OncoGel, Onxal, Oraxol, PTX, Pacitaxel, Pacliex, Padexol, Paxene, Plaxicel, QW 8184, Sindaxel, TAXUS Express, TAXUS stent, TaxAlbin, Taxol, Taxol A, Taxus, Taxus Liberte, Tocosol Paclitaxel, Yewtaxan, Zisu
InChI Formula InChI=1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32-,33+,35-,36+,37+,38-,40-,45+,46-,47+/m0/s1
SMILES CC(=O)O[C@H]1C(=O)[C@]2(C)[C@@H](O)C[C@H]3OC[C@@]3(OC(=O)C)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7)C(=C1C5(C)C)C
IUPAC Name 5β,20-Epoxy-1,7β-dihydroxy-9-oxotax-11-ene-2α,4,10β,13α-tetrayl 4,10-diacetate 2-benzoate 13-[(2R,3S)-3-(benzoylamino)-2-hydroxy-3-phenylpropanoate]
Molecular Weight (g/mol) 853.91
Formula Weight 853.331
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.