missing translation for 'onlineSavingsMsg'
Learn More

Paclitaxel, 99.5+%, Thermo Scientific™

Paclitaxel (CAS # 33069-62-4) is a microtubule inhibitor produced from the extract of the Pacific yew tree.

Brand:  Thermo Scientific Alfa Aesar J62734.03

Quantity 1g

 View more versions of this product

Product Code. 15485039

  • £573.00 / 1g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • Paclitaxel is a tetracyclic diterpenoid isolated from the Pacific yew, Taxus brevifolia
  • Paclitaxel acts as an antineoplastic agent by binding tubulin and preventing cell division
  • Paclitaxel may also bind Bcl-2 and lead to disinhibition of apoptosis

Application

  • Paclitaxel is used in the study of lung, breast, and ovarian cancers; it may be useful in the research of other cancer types
  • It is suitable for cell culture, biochemistry, and molecular biology applications
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

33069-62-4
853.92
RCINICONZNJXQF-VAZQATRQSA-N
133640187
CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1
C47H51NO14
MFCD00869953
paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel
(1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate

Specifications

33069-62-4
MFCD00869953
1420457
paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel
RCINICONZNJXQF-VAZQATRQSA-N
(1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
133640187
≥99.5%
C47H51NO14
1g
14,6982
Soluble in DMSO (200mg/ml) or ethanol (40mg/ml); Soluble in dimethylsulfoxide,methanol,ethanol,acetonitrile,cremophor and dimehtylsulfamide; Sparingly soluble in aqueous buffer.
CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1
853.92
853.91
Paclitaxel
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.