missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Paclitaxel

Product Code. 17150202
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17150202 25 mg 25mg
17160202 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17150202 Supplier STEMCELL Technologies Supplier No. 73312
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibitor of microtubule formation

  • Paclitaxel is a diterpene alkaloid originally isolated from the bark of the Pacific Yew tree (Taxus brevifolia)
  • It binds to and stabilizes microtubules, preventing their reorganization during cell division, which leads to cell cycle arrest
  • Paclitaxel has antitumorigenic properties and has been used as a chemotherapeutic compound (Rowinsky et al.) Many pathways have been implicated in Paclitaxel-induced apoptosis, including c-Jun N-terminal kinase/stress-activated protein kinase (JNK/SAPK), p38 mitogen-activated protein kinase (MAPK), and protein kinase A (PKA; Wang et al.; Reshkin et al.)

Specifications

CAS 33069-62-4
Synonym NSC 125973
Quantity 25 mg
Molecular Formula C47H51NO14
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.