Learn More
Restek Packed Column Molesieve 13X 60/80 6' 1/8" SS OC Var
Brand:  Restek 80475-820
Encompass_Preferred
Packed Column Molesieve 13X 60/80 6' 1/8" SS OC Var
Product Code. 16659916
  • £381.15 / Each