Learn More
Restek Packed Column 0.2% Carbowax 1500 80/100 CBK C 2m 1/8" OD 2mmID Silco Shimadzu 8A
Brand:  Restek 80122-851
Encompass_Preferred
Packed Column 0.2% Carbowax 1500 80/100 CBK C 2m 1/8" OD 2mmID Silco Shimadzu 8A
Product Code. 16659696
  • £429.55 / Each