Learn More
Thermo Scientific™ Cell Locker™ Chamber with Copper Tray and Transport Cover
Product Code. 15804295 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Places:
6
Unit Size:
Each
Product Code. No. of Places unitSize
15804295 6 Each
1 options
Product Code. 15804295 Supplier Thermo Scientific™ Supplier No. 50154739X6
Protect your most sensitive cells, even during transport.
Each individual Cell Locker™ Chamber is effectively a quarantine chamber. Can be manually disinfected or can be autoclaved.
  • Dual 0.2 μm membrane filters permit air circulation but exclude microbial contaminants
  • Replacable 0.2 μm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity
  • Dishes and flasks can be removed individually on a work tray or inside the entire chamber with optional transport cover, thus protecting samples from outside air
  • Non-skid tray slides out to allow access to culture vessels, or the entire tray can be removed
TRUSTED_SUSTAINABILITY
Specifications
No. of Places 6
For Use With (Application) Cell Protection and Transport
Includes Chamber, Copper Tray, and Transport Cover
Quantity 1 Ea.