missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Pacific Blue™ C5-Maleimide
Description
Pacific Blue™ C5-maleimide is an excellent reagent for thiol-selective modification, quantitation and analysis. In this reaction, the thiol is added across the double bond of the maleimide to yield a thioether. This compound does not react with methionine, histidine or tyrosine. Reaction of Pacific Blue™ C5-maleimide with amines usually requires a higher pH than reaction of maleimides with thiols.
View all Pacific Blue™ dye products..
View the Fluorophore Selection Guide.
View all Pacific Blue™ dye products..
View the Fluorophore Selection Guide.
Specifications
Specifications
| Format | Solid |
| Quantity | 1 mg |
| Product Type | Maleimide |
| Solubility | DMSO (Dimethylsulfoxide) |
| Content And Storage | Contains 1 vial of Pacific Blue™ dye (1 mg). Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Thiol |
| Detection Method | Fluorescence |
| Flow Cytometer Laser Lines | 405 |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Maleimide |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?