missing translation for 'onlineSavingsMsg'
Learn More

Pachymic acid, MedChemExpress

Product Code. 16458958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458958 10 mg 10mg
16451388 5 mg 5mg
16404827 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16458958 Supplier Medchem Express Supplier No. HYN0371
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways.

Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways.

Specifications

Chemical Name or Material Pachymic acid
CAS 29070-92-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H52O5
Quantity 10 mg
Synonym 3-O-Acetyltumulosic acid
Solubility Information DMSO : 10 mg/mL (18.91 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : 4.17 mg/mL (7.89 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O[C@@H]([C@]1([H])[C@H](C(O)=O)CCC(C(C)C)=C)C[C@@]([C@]1(C)CC2)(C)C3=C2[C@]4(C)[C@](CC3)([H])C(C)(C)[C@@H](OC(C)=O)CC4
Molecular Weight (g/mol) 528.76
Formula Weight 528.76
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.