missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PACAP-38 Antibody (Lilly patent anti-PACAP) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | PACAP-38 |
|---|---|
| Clone | Lilly patent anti-PACAP |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30047115
|
Novus Biologicals™
NBP3-28393-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046976
|
Novus Biologicals™
NBP3-28393-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047437
|
Novus Biologicals™
NBP3-28393-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PACAP-38 Monoclonal antibody specifically detects PACAP-38 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| PACAP-38 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Human | |
| P18509 | |
| 116 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Lilly patent anti-PACAP | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| pituitary adenylate cyclase-activating polypeptide, adenylate cyclase activating polypeptide 1 (pituitary), PACAP | |
| PACAP38 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title