missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAC2 Antibody (OTI1E8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73215CL1
This item is not returnable.
View return policy
Description
PAC2 Monoclonal antibody specifically detects PAC2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PAC2 | |
| Monoclonal | |
| CoraFluor 1 | |
| CLAST3HSPC260, HCCA3PAC-2, hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-susceptibility protein 3, HsT1707, likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3), MDS003, MGC15092, PAC2HDCMC29P, proteasome (prosome, macropain) assembly chaperone 2, proteasome assembling chaperone 2, proteasome assembly chaperone 2, TNFSF5IP1CD40 ligand-activated specific transcript 3, Tumor necrosis factor superfamily member 5-induced protein 1, tumor necrosis factor superfamily, member 5-induced protein 1, x 003 protein | |
| Full length human recombinant protein of human PSMG2 (NP_064617) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI1E8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 56984 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction