missing translation for 'onlineSavingsMsg'
Learn More

PAC-1, TRC

Product Code. 30420214
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420214 10 mg 10mg
30414527 5 mg 5mg
30415300 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420214 Supplier Toronto Research Chemicals Supplier No. TRCP13200010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 315183-21-2
Molecular Formula C23H28N4O2
Molecular Weight (g/mol) 392.49
Synonym 4-(Phenylmethyl)-1-piperazineacetic Acid 2-[[2-Hydroxy-3-(2-propen-1-yl)phenyl] methylene]hydrazide, Procaspase Activating Compound 1
IUPAC Name 2-(4-benzylpiperazin-1-yl)-N-[(E)-(2-hydroxy-3-prop-2-enylphenyl)methylideneamino]acetamide
SMILES Oc1c(CC=C)cccc1\C=N\NC(=O)CN2CCN(Cc3ccccc3)CC2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H28N4O2/c1-2-7-20-10-6-11-21(23(20)29)16-24-25-22(28)18-27-14-12-26(13-15-27)17-19-8-4-3-5-9-19/h2-6,8-11,16,29H,1,7,12-15,17-18H2,(H,25,28)/b24-16+
Formula Weight 392.2212
Chemical Name or Material PAC-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.