missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPN1 Rabbit anti-Human, Mouse, Rat, Clone: 7V4X8, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £376.00
Specifications
| Antigen | PABPN1 |
|---|---|
| Clone | 7V4X8 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18357114
|
Novus Biologicals
NBP3-15539-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18354694
|
Novus Biologicals
NBP3-15539-100UL |
100 μg |
£376.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PABPN1 Monoclonal antibody specifically detects PABPN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PABPN1 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Nuclear poly(A)-binding protein 1, OPMD, PAB2poly(A) binding protein II, PABII, PABP-2, PABP2polyadenylate-binding protein 2, poly(A) binding protein 2, poly(A) binding protein, nuclear 1, Poly(A)-binding protein 2, Poly(A)-binding protein II, poly(A)-binding protein, nuclear 1, Polyadenylate-binding nuclear protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PABPN1 (Q86U42). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7V4X8 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8106 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title