missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPN1 Antibody (6C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26444-100ul
This item is not returnable.
View return policy
Description
PABPN1 Monoclonal antibody specifically detects PABPN1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| PABPN1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 6C3 | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Nuclear poly(A)-binding protein 1, OPMD, PAB2poly(A) binding protein II, PABII, PABP-2, PABP2polyadenylate-binding protein 2, poly(A) binding protein 2, poly(A) binding protein, nuclear 1, Poly(A)-binding protein 2, Poly(A)-binding protein II, poly(A)-binding protein, nuclear 1, Polyadenylate-binding nuclear protein 1 | |
| A synthesized peptide derived from Human PABPN1 [UniProt Q86U42] | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 8106 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction