missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PABPC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551358
This item is not returnable.
View return policy
Description
Immunogen sequence: AEVRTRDRAT FTNVFVKNIG DDIDDEKLKE LFCEYGPTES VKVIRDASGK SKGFGFVRYE THEAAQKAVL DLHGKSIDGK VLYVGRAQKK IERLAELRRR FERLRL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
PABPC5 encodes a protein that binds to the polyA tail found at the 3' end of most eukaryotic mRNAs. It is thought to play a role in the regulation of mRNA metabolic processes in the cytoplasm. PABPC5 is located in a gene-poor region within the X-specific 13d-sY43 subinterval of the chromosome Xq21.3/Yp11.2 homology block. It is located close to translocation breakpoints associated with premature ovarian failure, and is therefore a potential candidate gene for this disorder.
Specifications
| PABPC5 | |
| Polyclonal | |
| Unconjugated | |
| PABPC5 | |
| C820015E17; PABP5; PABP-5; Pabpc5; poly A binding protein, cytoplasmic 5; poly A binding protein, cytoplasmic 5-like; poly(A) binding protein cytoplasmic 5; poly(A) binding protein, cytoplasmic 5; polyadenylate-binding protein 5; polyadenylate-binding protein 5-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96DU9 | |
| PABPC5 | |
| Recombinant protein corresponding to Human PABPC5. Recombinant protein control fragment (Product #RP-91973). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction