missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19672
This item is not returnable.
View return policy
Description
PABPC5 Polyclonal specifically detects PABPC5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PABPC5 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| cytoplasmic 5, FLJ51677, PABP-5, poly(A) binding protein, cytoplasmic 5, Poly(A)-binding protein 5 | |
| Rabbit | |
| 43 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat. Species reactivity based on sequence identity: Rhesus Monkey (100%), Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q96DU9 | |
| PABPC5 | |
| Recombinant protein encompassing a sequence within the center region of human PABPC5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 140886 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction