missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PABPC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555575
This item is not returnable.
View return policy
Description
Immunogen sequence: RPQGFQGMPS AIRQSGPRPT LRHLAPTGSE CPDRLAMDFG GAGAAQQGLT DSCQSGGVPT AVQNLAPRAA VAAAAP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Poly(A)-binding proteins (PABPs) bind to the poly(A) tail present at the 3-prime ends of most eukaryotic mRNAs. PABPC4 or IPABP (inducible PABP) was isolated as an activation-induced T-cell mRNA encoding a protein. Activation of T cells increased PABPC4 mRNA levels in T cells approximately 5-fold. PABPC4 contains 4 RNA-binding domains and proline-rich C terminus. PABPC4 is localized primarily to the cytoplasm. It is suggested that PABPC4 might be necessary for regulation of stability of labile mRNA species in activated T cells. PABPC4 was also identified as an antigen, APP1 (activated-platelet protein-1), expressed on thrombin-activated rabbit platelets. PABPC4 may also be involved in the regulation of protein translation in platelets and megakaryocytes or may participate in the binding or stabilization of polyadenylates in platelet dense granules. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PABPC4 | |
| Polyclonal | |
| Unconjugated | |
| PABPC4 | |
| Activated-platelet protein 1; APP1; APP-1; Inducible poly(A)-binding protein; iPABP; PABP4; PABP-4; PABPC4; poly(A) binding protein cytoplasmic 4; poly(A) binding protein, cytoplasmic 4; poly(A) binding protein, cytoplasmic 4 (inducible form); poly(A)-binding protein 4; polyadenylate-binding protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8761 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13310 | |
| PABPC4 | |
| Recombinant protein corresponding to Human PABPC4. Recombinant protein control fragment (Product #RP-94047). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction