missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-74594
This item is not returnable.
View return policy
Description
PABPC4 Polyclonal specifically detects PABPC4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| PABPC4 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| APP1FLJ43938, APP-1PABP-4, Inducible poly(A)-binding protein, IPABP, iPABPpoly(A)-binding protein, cytoplasmic 4 (inducible form), PABP4Activated-platelet protein 1, poly(A) binding protein, cytoplasmic 4 (inducible form), Poly(A)-binding protein 4, polyadenylate-binding protein 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8761 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 2-5ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Q13310 | |
| PABPC4 | |
| The immunogen recognized by this antibody maps to a region between residue 594 and 644 of human Poly(A)-binding protein 4 using the numbering given in entry NP_003810.1 (GeneID 8761). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction