missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15061
This item is not returnable.
View return policy
Description
PABPC3 Polyclonal specifically detects PABPC3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| PABPC3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| PABP-3, PABP3polyadenylate-binding protein 3, PABPL3testis PABP, poly(A) binding protein, cytoplasmic 3, Poly(A)-binding protein 3, poly(A)-binding protein, cytoplasmic 3, poly(A)-binding protein-like 3, Testis-specific poly(A)-binding protein, testis-specific poly(A)-binding protein (PABP), testis-specific poly(A)-binding protein 3, tPABP | |
| Rabbit | |
| 70 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9H361 | |
| PABPC3 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PABPC3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5042 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction