missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | PABPC1L |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18237604
|
Novus Biologicals
NBP2-55978 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645398
|
Novus Biologicals
NBP2-55978-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PABPC1L Polyclonal specifically detects PABPC1L in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PABPC1L | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 80336 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ILTNQYMQRLSTMRTLSNPLLGSFQQPSSYFLPAMPQPPAQAAYYGCGPVTPTQP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| C20orf119, Chromosome 20 Open Reading Frame 119, DJ1069P2.3, Embryonic Poly(A) Binding Protein, EPAB, PABPC1L1, Poly(A) Binding Protein Cytoplasmic 1 Like, Poly(A) Binding Protein, Cytoplasmic 1-Like, Polyadenylate-Binding Protein 1-Like | |
| PABPC1L | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title