missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PABP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517599
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4), and PABP3 (PABPC3). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.
Specifications
| PABP | |
| Polyclonal | |
| Unconjugated | |
| PABPC1 | |
| Cytoplasmic poly(A)-binding protein 1-A; ePAB; hypothetical protein; PAB1; pabp; PABP 1; Pabp1; PABP-1; PABP-1A; PABP-1-A; PABP-A; Pabpc1; pabpc1.L; pabpc1.S; pabpc1a; pabpc1-a; pabpc1a protein; pabpc1-b; pabpc2; PabpI; PABPL1; poly A binding protein 1; poly A binding protein, cytoplasmic 1 a; poly(A) binding protein cytoplasmic 1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 1 L homeolog; poly(A) binding protein, cytoplasmic 1 S homeolog; poly(A) binding protein, cytoplasmic 1a; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1; poly(A)-binding protein 1A; poly(A)-binding protein 1-A; polyadenylate-binding protein 1; Polyadenylate-binding protein 1A; Polyadenylate-binding protein 1-A; polyadenylate-binding protein 1A; poly(A) binding protein, cytoplasmic 1a; wu:fb16a02; wu:fi19b08; wu:fj12d09; wu:fj61f06; XELAEV_18033915mg; xPABP1-A; zgc:109879 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 171350, 18458, 26986 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, RNA Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 74 μg/mL | |
| 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
| P11940, P29341, Q9EPH8 | |
| PABPC1 | |
| Synthetic peptide corresponding to amino acids at the amino-terminus of human PABP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction