missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PABP Monoclonal antibody specifically detects PABP in Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | PABP |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 1G1 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation |
| Formulation | Supplied as concentrated hybridoma cell culture media. |
| Gene Alias | PAB1polyadenylate-binding protein 1, PABP, PABP-1, PABP1poly(A) binding protein, cytoplasmic 2, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 1, Poly(A)-binding protein 1, poly(A)-binding protein, cytoplasmic 2 |
| Host Species | Mouse |
| Immunogen | Yeast polyA binding protein preparations. UniProt: P04147 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?