missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PABP Monoclonal Antibody (10E10)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA134079

Product Code. 11594063

  • £514.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-34079 detects PABP from human, chicken, rabbit and Xenopus laevis samples. Does not react wtih Drosophilia melanogaster or mouse samples.

The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4), and PABP3 (PABPC3). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PABP
Monoclonal
1.0 mg/mL
PBS with 0.1% sodium azide
P11940, P20965
PABPC1, pabpc1.S
Recombinant PABP (Human) expressed from its 1.85 kbp cDNA, NcoI to SspI.
50 μL
Primary
Human, Rabbit, Chicken, Frog
Antibody
IgG2b
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
10E10
Unconjugated
PABPC1
Cytoplasmic poly(A)-binding protein 1-A; ePAB; hypothetical protein; PAB1; pabp; PABP 1; Pabp1; PABP-1; PABP-1A; PABP-1-A; PABP-A; Pabpc1; pabpc1.L; pabpc1.S; pabpc1a; pabpc1-a; pabpc1a protein; pabpc1-b; pabpc2; PabpI; PABPL1; poly A binding protein 1; poly A binding protein, cytoplasmic 1 a; poly(A) binding protein cytoplasmic 1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 1 L homeolog; poly(A) binding protein, cytoplasmic 1 S homeolog; poly(A) binding protein, cytoplasmic 1a; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1; poly(A)-binding protein 1A; poly(A)-binding protein 1-A; polyadenylate-binding protein 1; Polyadenylate-binding protein 1A; Polyadenylate-binding protein 1-A; polyadenylate-binding protein 1A; poly(A) binding protein, cytoplasmic 1a; wu:fb16a02; wu:fi19b08; wu:fj12d09; wu:fj61f06; XELAEV_18033915mg; xPABP1-A; zgc:109879
Mouse
Protein A
RUO
100342434, 26986, 379896, 430997
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.