missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABP Antibody (10E10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80901
This item is not returnable.
View return policy
Description
PABP Monoclonal specifically detects PABP in Human, Bovine, Canine, Primate, Xenopus, Drosophila (Negative), Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray, CyTOF-ready.
Specifications
| PABP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry, ELISA : 1:100-1:2000, Immunocytochemistry/Immunofluorescence : 1:50-1:100, Immunoprecipitation : 1:10-1:500, Microarray, CyTOF-ready | |
| PAB1polyadenylate-binding protein 1, PABP, PABP-1, PABP1poly(A) binding protein, cytoplasmic 2, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 1, Poly(A)-binding protein 1, poly(A)-binding protein, cytoplasmic 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Monkey, Bovine, Dog, and Hamster and Xenopus laevis. Does not cross-react with Drosophilia melanogaster or Mouse. Not yet tested in other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, CyTOF | |
| 1e+011 | |
| Unconjugated | |
| PBS | |
| PABPC1 | |
| Recombinant human PABP protein. [Uniprot# P11940] | |
| 0.1 mL | |
| Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| 26986 | |
| Human, Bovine, Canine, Primate, Xenopus species, Drosophila (Negative), Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction