missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA2G4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15348-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against full length PA2G4 of human origin.
This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.Specifications
| PA2G4 | |
| Polyclonal | |
| Unconjugated | |
| PA2G4 | |
| EBP1, ErbB3 binding protein 1, HG4 1, p38 2G4, PA2G4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18813, 288778, 5036 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50580, Q6AYD3, Q9UQ80 | |
| Pa2g4 | |
| PA2G4 Fusion Protein Ag7587 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur