missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA2G4/EBP1 Antibody (OTI1D3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73213CL1
This item is not returnable.
View return policy
Description
PA2G4/EBP1 Monoclonal antibody specifically detects PA2G4/EBP1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PA2G4/EBP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, hG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4 | |
| Full length human recombinant protein of human PA2G4 (NP_006182) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 5036 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| OTI1D3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction